Assay Details
Gene Symbol: Slc3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271089.2 | NP_001258018.1 | 8 - 9 | 1313 | 86 | |
NM_019283.3 | NP_062156.2 | 9 - 10 | 1438 | 86 | ||
GenBank mRNA | AB015433.1 | - | 9 - 10 | 1404 | 86 | |
BC061989.1 | - | 8 - 9 | 1296 | 86 | ||
U59324.1 | - | 8 - 9 | 1227 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
