Assay Details
Gene Symbol: Frmd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008348.1 | NP_001008349.1 | 1 - 2 | 159 | 56 | |
XM_006230804.2 | XP_006230866.1 | 1 - 2 | 186 | 56 | ||
GenBank mRNA | BC085896.1 | - | 1 - 2 | 159 | 7193163 | 56 |
FQ211651.1 | - | 1 - 2 | 198 | 56 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC085896.1, NM_001008348.1, XM_006230804.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
