Assay Details
Gene Symbol: Frmd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008348.1 | NP_001008349.1 | 2 - 3 | 244 | 131 | |
GenBank mRNA | BC085896.1 | - | 2 - 3 | 244 | 131 | |
FQ211651.1 | - | 2 - 3 | 283 | 131 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC085896.1, NM_001008348.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
