Assay Details
Gene Symbol: mrpl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006973.1 | NP_001006974.1 | 3 - 4 | 352 | 86 | |
XM_006230739.2 | XP_006230801.1 | 3 - 4 | 396 | 86 | ||
GenBank mRNA | BC083740.1 | - | 3 - 4 | 352 | 7191782 | 86 |
FQ225172.1 | - | 3 - 4 | 377 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
