Assay Details
Gene Symbol: Frs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108097.3 | NP_001101567.2 | 3 - 4 | 300 | 118 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108097.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
