Assay Details
Gene Symbol: Gns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011989.1 | NP_001011989.1 | 1 - 2 | 186 | 87 | |
XM_006241396.2 | XP_006241458.1 | 1 - 2 | 282 | 87 | ||
GenBank mRNA | BC087741.1 | - | 1 - 2 | 186 | 87 |
Gene Symbol: LOC100909505 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750325.3 | XP_003750373.1 | 283 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
