Assay Details
Gene Symbol: Irak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108101.1 | NP_001101571.1 | 4 - 5 | 614 | 124 | |
GenBank mRNA | BC166731.1 | - | 4 - 5 | 689 | 7934689 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
