Assay Details
Gene Symbol: Irak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108101.1 | NP_001101571.1 | 8 - 9 | 1066 | 87 | |
GenBank mRNA | BC166731.1 | - | 8 - 9 | 1141 | 7934689 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
