Assay Details
Gene Symbol: Etf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008344.1 | NP_001008345.1 | 7 - 8 | 961 | 81 | |
GenBank mRNA | BC085902.1 | - | 7 - 8 | 961 | 7193574 | 81 |
FQ227237.1 | - | 7 - 8 | 990 | 81 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
