Assay Details
Gene Symbol: Msl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014111.1 | NP_001014133.1 | 4 - 5 | 392 | 69 | |
XM_006256835.2 | XP_006256897.2 | 5 - 6 | 778 | 69 | ||
XM_006256836.2 | XP_006256898.2 | 5 - 6 | 781 | 69 | ||
XM_006256839.2 | XP_006256901.2 | 5 - 6 | 778 | 69 | ||
XM_008773169.1 | XP_008771391.1 | 5 - 6 | 781 | 69 | ||
GenBank mRNA | BC086548.1 | - | 4 - 5 | 392 | 69 |
Gene Symbol: LOC102547271 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256847.2 | XP_006256909.1 | 1 - 2 | 104 | 69 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_006256847.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
