Assay Details
Gene Symbol: Cdip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008360.1 | NP_001008361.1 | 5 - 6 | 499 | 87 | |
XM_006245811.2 | XP_006245873.1 | 4 - 5 | 510 | 87 | ||
XM_006245812.2 | XP_006245874.1 | 6 - 7 | 697 | 87 | ||
XM_006245813.2 | XP_006245875.1 | 5 - 6 | 1020 | 87 | ||
XM_006245814.2 | XP_006245876.1 | 5 - 6 | 623 | 87 | ||
XM_006245815.2 | XP_006245877.1 | 4 - 5 | 946 | 87 | ||
XM_006245816.2 | XP_006245878.1 | 4 - 5 | 513 | 87 | ||
XM_008767493.1 | XP_008765715.1 | 4 - 5 | 1924 | 87 | ||
XM_008767494.1 | XP_008765716.1 | 4 - 5 | 2175 | 87 | ||
GenBank mRNA | BC085860.1 | - | 5 - 6 | 499 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
