Assay Details
Gene Symbol: Eef2kmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106975.1 | NP_001100445.1 | 2 - 3 | 175 | 69 | |
XM_006245785.2 | XP_006245847.1 | 2 - 3 | 243 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
