Assay Details
Gene Symbol: Ufm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126080.2 | NP_001119552.1 | 4 - 5 | 241 | 94 | |
GenBank mRNA | BC091395.1 | - | 4 - 5 | 177 | 94 | |
CB787557.1 | - | 4 - 5 | 279 | 94 | ||
CV112030.1 | - | 3 - 4 | 131 | 94 | ||
FQ229705.1 | - | 4 - 5 | 212 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
