Assay Details
Gene Symbol: Slc1a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108973.1 | NP_001102443.1 | 7 - 8 | 1280 | 74 | |
XM_008763892.1 | XP_008762114.1 | 7 - 8 | 1209 | 74 | ||
GenBank mRNA | JF422064.1 | - | 7 - 8 | 917 | 74 | |
JF422065.1 | - | 6 - 7 | 866 | 74 | ||
JF422066.1 | - | 7 - 8 | 1082 | 74 | ||
JF422068.1 | - | 7 - 8 | 1082 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
