Assay Details
Gene Symbol: Urod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019209.1 | NP_062082.1 | 8 - 9 | 911 | 73 | |
GenBank mRNA | BC158690.1 | - | 8 - 9 | 911 | 8376316 | 73 |
BI282626.1 | - | 2 - 3 | 73 | |||
FQ216026.1 | - | 8 - 9 | 906 | 73 | ||
FQ216320.1 | - | 8 - 9 | 887 | 73 | ||
FQ218910.1 | - | 8 - 9 | 902 | 73 | ||
FQ230458.1 | - | 9 - 10 | 756 | 73 | ||
FQ233169.1 | - | 8 - 9 | 888 | 73 | ||
FQ234620.1 | - | 8 - 9 | 909 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
