Assay Details
Gene Symbol: Eri3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238735.2 | XP_006238797.1 | 2 - 3 | 439 | 74 | |
XM_006238739.1 | XP_006238801.1 | 2 - 3 | 439 | 74 | ||
XM_006238741.2 | XP_006238803.1 | 2 - 3 | 135 | 74 | ||
XM_008764085.1 | XP_008762307.1 | 2 - 3 | 2458 | 74 | ||
XM_233435.6 | XP_233435.5 | 2 - 3 | 439 | 74 | ||
GenBank mRNA | BC167080.1 | - | 1 - 2 | 79 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006238741.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
