Assay Details
Gene Symbol: Eri3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238735.2 | XP_006238797.1 | 4 - 5 | 836 | 74 | |
XM_006238736.2 | XP_006238798.1 | 4 - 5 | 830 | 74 | ||
XM_006238737.2 | XP_006238799.1 | 5 - 6 | 645 | 74 | ||
XM_006238738.2 | XP_006238800.1 | 3 - 4 | 2887 | 74 | ||
XM_006238739.1 | XP_006238801.1 | 4 - 5 | 836 | 74 | ||
XM_006238740.2 | XP_006238802.1 | 3 - 4 | 252 | 74 | ||
XM_006238741.2 | XP_006238803.1 | 4 - 5 | 532 | 74 | ||
XM_008764085.1 | XP_008762307.1 | 4 - 5 | 2855 | 74 | ||
XM_008764086.1 | XP_008762308.1 | 3 - 4 | 831 | 74 | ||
XM_233435.6 | XP_233435.5 | 4 - 5 | 836 | 74 | ||
GenBank mRNA | BC167080.1 | - | 3 - 4 | 476 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
