Assay Details
Gene Symbol: Acvr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199230.1 | NP_954700.1 | 7 - 8 | 1270 | 94 | |
XM_006242315.2 | XP_006242377.1 | 7 - 8 | 2114 | 94 | ||
XM_008765678.1 | XP_008763900.1 | 7 - 8 | 5275 | 94 | ||
GenBank mRNA | S76466.1 | - | 7 - 8 | 1270 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
