Assay Details
Gene Symbol: Racgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108112.1 | NP_001101582.1 | 13 - 14 | 1630 | 79 | |
XM_006257400.2 | XP_006257462.1 | 13 - 14 | 1577 | 79 | ||
GenBank mRNA | BC166473.1 | - | 13 - 14 | 1530 | 9090730 | 79 |
FQ231766.1 | - | 5 - 6 | 667 | 79 | ||
FQ234782.1 | - | 5 - 6 | 667 | 79 |
Gene Symbol: LOC103690014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765882.1 | XP_008764104.1 | 11 - 12 | 1206 | 79 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
