Assay Details
Gene Symbol: Racgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108112.1 | NP_001101582.1 | 15 - 16 | 1880 | 62 | |
XM_006257400.2 | XP_006257462.1 | 15 - 16 | 1827 | 62 | ||
GenBank mRNA | BC166473.1 | - | 15 - 16 | 1780 | 62 | |
FQ234782.1 | - | 7 - 8 | 917 | 62 |
Gene Symbol: LOC103690014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765882.1 | XP_008764104.1 | 13 - 14 | 1456 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
