Assay Details
Gene Symbol: Ranbp3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232030.2 | XP_006232092.1 | 813 | 73 | ||
XM_006232031.2 | XP_006232093.1 | 737 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
