Assay Details
Gene Symbol: Elmod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109506.1 | NP_001102976.1 | 1 - 2 | 101 | 145 | |
XM_006255303.2 | XP_006255365.1 | 1 - 2 | 147 | 145 |
Gene Symbol: LOC102554034 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772180.1 | XP_008770402.1 | 1 | 72 | 145 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109506.1, XM_006255303.2, XM_008772180.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
