Assay Details
Gene Symbol: Elmod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109506.1 | NP_001102976.1 | 5 - 6 | 499 | 91 | |
XM_006255302.2 | XP_006255364.1 | 5 - 6 | 746 | 91 | ||
XM_006255303.2 | XP_006255365.1 | 5 - 6 | 545 | 91 |
Gene Symbol: LOC102554034 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772180.1 | XP_008770402.1 | 1 | 470 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
