Assay Details
Gene Symbol: E2f4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271345.1 | NP_001258274.1 | 5 - 6 | 597 | 129 | |
XM_006255433.1 | XP_006255495.1 | 5 - 6 | 592 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
