Assay Details
Gene Symbol: Usp38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751862.3 | XP_003751910.1 | 2 - 3 | 1349 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
