Assay Details
Gene Symbol: Usp38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751862.3 | XP_003751910.1 | 4 - 5 | 1581 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
