Assay Details
Gene Symbol: Ppm1j | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005540.1 | NP_001005540.1 | 2 - 3 | 447 | 59 | |
XM_008761380.1 | XP_008759602.1 | 3 - 4 | 945 | 59 | ||
XM_008761381.1 | XP_008759603.1 | 3 - 4 | 945 | 59 | ||
GenBank mRNA | BC082053.1 | - | 2 - 3 | 447 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
