Assay Details
Gene Symbol: Pip5k1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042621.1 | NP_001036086.1 | 13 - 14 | 1918 | 70 | |
XM_008761315.1 | XP_008759537.1 | 13 - 14 | 2047 | 70 | ||
XM_008761316.1 | XP_008759538.1 | 13 - 14 | 2044 | 70 | ||
GenBank mRNA | AY724476.1 | - | 13 - 14 | 1918 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
