Assay Details
Gene Symbol: Slc22a15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107707.1 | NP_001101177.1 | 7 - 8 | 1208 | 81 | |
XM_006233050.2 | XP_006233112.1 | 6 - 7 | 3602 | 81 | ||
GenBank mRNA | FQ211559.1 | - | 3 - 4 | 789 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
