Assay Details
Gene Symbol: Mtmr9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005761.1 | NP_001005761.1 | 5 - 6 | 1042 | 79 | |
GenBank mRNA | AF526269.1 | - | 5 - 6 | 654 | 79 | |
BC083644.1 | - | 5 - 6 | 1042 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
