Assay Details
Gene Symbol: Blk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025751.1 | NP_001020922.1 | 1 - 2 | 355 | 82 | |
GenBank mRNA | BC098683.1 | - | 1 - 2 | 355 | 82 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC098683.1, NM_001025751.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
