Assay Details
Gene Symbol: Blk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025751.1 | NP_001020922.1 | 6 - 7 | 811 | 77 | |
XM_006252188.2 | XP_006252250.1 | 7 - 8 | 1205 | 77 | ||
XM_008770769.1 | XP_008768991.1 | 6 - 7 | 1232 | 77 | ||
XM_008770770.1 | XP_008768992.1 | 5 - 6 | 471 | 77 | ||
GenBank mRNA | BC098683.1 | - | 6 - 7 | 811 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
