Assay Details
Gene Symbol: Blk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025751.1 | NP_001020922.1 | 9 - 10 | 1283 | 69 | |
XM_006252188.2 | XP_006252250.1 | 10 - 11 | 1677 | 69 | ||
XM_008770769.1 | XP_008768991.1 | 9 - 10 | 1704 | 69 | ||
XM_008770770.1 | XP_008768992.1 | 8 - 9 | 943 | 69 | ||
GenBank mRNA | BC098683.1 | - | 9 - 10 | 1283 | 7385175 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
