Assay Details
Gene Symbol: Hibadh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022243.1 | NP_071579.1 | 1 - 2 | 122 | 124 | |
GenBank mRNA | BC127442.1 | - | 1 - 2 | 108 | 7308860 | 124 |
J04628.1 | - | 1 - 2 | 123 | 125 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC127442.1, NM_022243.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
