Assay Details
Gene Symbol: Hibadh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022243.1 | NP_071579.1 | 5 - 6 | 649 | 105 | |
GenBank mRNA | BC127442.1 | - | 5 - 6 | 635 | 7308860 | 105 |
J04628.1 | - | 5 - 6 | 650 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
