Assay Details
Gene Symbol: Atp6v1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108318.2 | NP_001101788.1 | 3 - 4 | 316 | 95 | |
XM_006248348.2 | XP_006248410.1 | 3 - 4 | 517 | 95 | ||
XM_006248349.2 | XP_006248411.1 | 3 - 4 | 508 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
