Assay Details
Gene Symbol: Atp6v1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108318.2 | NP_001101788.1 | 4 - 5 | 532 | 129 | |
XM_006248348.2 | XP_006248410.1 | 4 - 5 | 733 | 129 | ||
XM_006248349.2 | XP_006248411.1 | 4 - 5 | 724 | 129 | ||
GenBank mRNA | FQ211993.1 | - | 4 - 5 | 565 | 129 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
