Assay Details
Gene Symbol: Atp6v1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108318.2 | NP_001101788.1 | 6 - 7 | 820 | 67 | |
XM_006248348.2 | XP_006248410.1 | 6 - 7 | 1021 | 67 | ||
XM_006248349.2 | XP_006248411.1 | 6 - 7 | 1012 | 67 | ||
GenBank mRNA | FQ211993.1 | - | 6 - 7 | 853 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
