Assay Details
Gene Symbol: Ift57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107093.1 | NP_001100563.1 | 10 - 11 | 1156 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
neural tube closure
heart looping
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
smoothened signaling pathway
intraciliary transport
regulation of apoptotic process
negative regulation of epithelial cell proliferation
cilium assembly
left/right pattern formation
neural tube closure
heart looping
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
smoothened signaling pathway
intraciliary transport
regulation of apoptotic process
negative regulation of epithelial cell proliferation
cilium assembly
left/right pattern formation
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P18356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529727 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67009 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741524 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745445 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924693 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657611 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756554 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895870 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738312 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165467 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P18356 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P18356 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P18356 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P18356 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 102297 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 6942 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10753792 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753789 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753793 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753787 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753785 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753791 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753794 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753790 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753788 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753786 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10753784 | - | - |
Rat Genome 230 2.0 Array | 1380548_at | - | - |
Rat Genome 230 2.0 Array | 1380548_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1380548_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1380548_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1100009195.rn.1 | TC1100001513.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1100009196.rn.1 | TC1100001513.rn.1 | RTA TC |
Rat Expression Set 230 | 1380548_PM_at | - | - |
Rat Expression Set 230 | 1380548_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10753784 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21565006 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351336 | - | - |
