Assay Details
Gene Symbol: Arhgap36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257554.2 | XP_006257616.2 | 10 - 11 | 1538 | 72 | |
XM_006257555.2 | XP_006257617.1 | 10 - 11 | 1561 | 72 | ||
XM_229133.8 | XP_229133.5 | 10 - 11 | 1538 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
