Assay Details
Gene Symbol: Arhgap36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257554.2 | XP_006257616.2 | 3 - 4 | 477 | 55 | |
XM_006257555.2 | XP_006257617.1 | 3 - 4 | 500 | 55 | ||
XM_229133.8 | XP_229133.5 | 3 - 4 | 477 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
