Assay Details
Gene Symbol: Arhgap36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257554.2 | XP_006257616.2 | 7 - 8 | 1106 | 93 | |
XM_006257555.2 | XP_006257617.1 | 7 - 8 | 1129 | 93 | ||
XM_229133.8 | XP_229133.5 | 7 - 8 | 1106 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
