Assay Details
Gene Symbol: Zfp280c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257541.2 | XP_006257603.1 | 399 | 140 | ||
XM_006257542.2 | XP_006257604.1 | 298 | 140 | |||
XM_008773539.1 | XP_008771761.1 | 4 - 5 | 286 | 140 | ||
XM_008773540.1 | XP_008771762.1 | 255 | 140 | |||
XM_008773541.1 | XP_008771763.1 | 3 - 4 | 250 | 140 | ||
XM_008773542.1 | XP_008771764.1 | 2 - 3 | 1280 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
