Assay Details
Gene Symbol: Slc35d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106668.2 | NP_001100138.1 | 2 - 3 | 190 | 73 | |
GenBank mRNA | FQ220185.1 | - | 2 - 3 | 167 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
