Assay Details
Gene Symbol: Itgb3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013213.1 | NP_001013231.1 | 5 - 6 | 657 | 90 | |
XM_008763882.1 | XP_008762104.1 | 3 - 4 | 474 | 90 | ||
XM_008763883.1 | XP_008762105.1 | 6 - 7 | 399 | 90 | ||
XM_008763884.1 | XP_008762106.1 | 5 - 6 | 353 | 90 | ||
GenBank mRNA | BC086363.1 | - | 5 - 6 | 657 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
