Assay Details
Gene Symbol: Ror1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108671.1 | NP_001102141.1 | 4 - 5 | 904 | 71 | |
XM_008763887.1 | XP_008762109.1 | 3 - 4 | 1754 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
