Assay Details
Gene Symbol: Arhgap35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271132.1 | NP_001258061.1 | 5 - 6 | 4726 | 72 | |
XM_006228412.2 | XP_006228474.1 | 5 - 6 | 5291 | 72 | ||
XM_008758918.1 | XP_008757140.1 | 6 - 7 | 4551 | 72 | ||
XM_008758919.1 | XP_008757141.1 | 5 - 6 | 4344 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
