Assay Details
Gene Symbol: Arhgap35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271132.1 | NP_001258061.1 | 6 - 7 | 4826 | 99 | |
XM_006228412.2 | XP_006228474.1 | 6 - 7 | 5391 | 99 | ||
XM_008758918.1 | XP_008757140.1 | 7 - 8 | 4651 | 99 | ||
XM_008758919.1 | XP_008757141.1 | 6 - 7 | 4444 | 99 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
