Assay Details
Gene Symbol: Samd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107498.1 | NP_001100968.1 | 4 - 5 | 1220 | 99 | |
XM_008759143.1 | XP_008757365.1 | 5 - 6 | 1343 | 99 | ||
GenBank mRNA | BC090319.1 | - | 5 - 6 | 1158 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
