Assay Details
Gene Symbol: Rinl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106241.1 | NP_001099711.1 | 10 - 11 | 1602 | 77 | |
XM_006228653.2 | XP_006228715.1 | 10 - 11 | 1854 | 77 | ||
XM_006228654.2 | XP_006228716.1 | 10 - 11 | 1842 | 77 | ||
XM_006228657.2 | XP_006228719.1 | 9 - 10 | 1577 | 77 | ||
XM_008759120.1 | XP_008757342.1 | 9 - 10 | 1719 | 77 | ||
XM_008759121.1 | XP_008757343.1 | 10 - 11 | 1687 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
